Learn More
Brady™ Thermal Transfer Printable Nylon Cloth Labels
Product Code. 10180213
Change view
Click to view available options
Size:
0.25 x 2 in. (L x W)
0.375 x 1 in. (L x W)
0.375 x 1 x 0.375 in. (L x W x dia.)
0.5 in. (dia.)
0.5 x 0.9 in. (L x W)
0.75 x 1.5 in. (L x W)
2 x 1 in. (L x W)
Quantity:
1500 Pk.
3000 Pk.
Unit Size:
1 roll
Pack of 1500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Quantity unitSize
10180213 0.375 x 1 x 0.375 in. (L x W x dia.) 3000 Pk. 1 roll
12869835 0.5 x 0.9 in. (L x W) 1500 Pk. 1 roll
15132243 2 x 1 in. (L x W) 1500 Pk. 1 roll
12624585 0.75 x 1.5 in. (L x W) 1500 Pk. 1 roll
15482143 0.375 x 1 in. (L x W) 1500 Pk. 1 roll
13089292 0.25 x 2 in. (L x W) 1500 Pk. 1 roll
30041427 0.5 in. (dia.) 1500 Pk. Pack of 1500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10180213 Supplier Brady™ Supplier No. 019260
Only null left
Add to basket
Add to basket
Thermal Transfer Printable Nylon Cloth Labels with high adhesion for use in autoclave, freezer or liquid nitrogen.
  • White, matte finished labels suitable for use in laboratory identifications and vial labeling.
  • Low profile.
  • Recommended for use with Brady thermal transfer printer ribbon series 4900.
  • Compatible with color Brady series 6200, 6000, 4300, and 4500.
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL, CSA C22.2 No.0.15-95, RoHS 2002/95/EC, DIN VDE 0472 part 815
Description Thermal Transfer Printer Labels
Adhesive Type Acrylic, Permanent
Color White
Finish Matte
Format Label
Material Nylon Cloth
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) For laboratory identification, vial top
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Includes Roll of 3000 Labels
Labels per Row 1
Life Span 2 Years
Min. Operating Temperature (Metric) -40°C
Quantity 3000 Pk.
Shape Rectangle with Rounded Corners
Size 0.375 x 1 x 0.375 in. (L x W x dia.)
Size (English) 1 x 0.375 in.
Size (Metric) 2.5 x 0.95 cm
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Tensile Strength 80 lb./in.
Thickness (English) 0.0065 in.
Thickness (Metric) 0.166 mm
Vertical Repeat (Metric) 12.70 mm
Dimensions (LxW) 25.40 mm (W) x 9.53 mm (H) x 9.53 mm (Dia)
Length (English) 0.37 in.
Width (English) 1 in.
Width (Metric) 25.40 mm
Width (English) Tape 1 in.
Width (Metric) Printable Area 42.55 mm
Height (English) 0.37 in.
Height (Metric) 9.53 mm
Diameter (English) 0.375 in.
Diameter (Metric) 9.53 mm
Alternate Part Number THT-163-499-3