Learn More
Brady™ BMP51/BMP53/BMP41 Label Maker Cartridge: B-422 Polyester
Click to view available options
Dimensions (LxW):
0.5 x 1.5 in.
1 x 0.5 in.
1 x 1 in.
1.5 x 0.75 in.
25 ft. x 0.475 in.
25 ft. x 0375 in.
25 ft. x 1 in.
Unit Size:
Each
Description
- Permanent adhesive, for powder-coated/textured surfaces
- Available in various sizes
Specifications
Specifications
| Certifications/Compliance | Pass 2002/95/EC |
| Color | Black, White |
| Material | Polyester (B-422) |
| Product Type | Label Maker Cartridge |
| Quantity | 260/Pk. |
| Size (English) | 1 x 1 in. |
| Size (Metric) | 25.4 x 25.4 mm |
| Dimensions (LxW) | 1 x 1 in. |
| Width (English) | 1 in. |
| Width (Metric) | 25.4 mm |
| Show More |