Learn More
Brady™ BMP51/BMP53/BMP41 Label Maker Cartridge: B-422 Polyester
Product Code. 15103329
Click to view available options
Dimensions (LxW):
0.5 x 1.5 in.
1 x 0.5 in.
1 x 1 in.
1.5 x 0.75 in.
25 ft. x 0.475 in.
25 ft. x 0375 in.
25 ft. x 1 in.
Unit Size:
Each
Product Code. 15103329
Brand: Brady™ 131592
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use in commercial and industrial applications
  • Permanent adhesive, for powder-coated/textured surfaces
  • Available in various sizes
Specifications
Certifications/Compliance Pass 2002/95/EC
Color Black, White
Material Polyester (B-422)
Product Type Label Maker Cartridge
Quantity 480/Pk.
Size (English) 1 x 0.5 in.
Size (Metric) 25.4 x 12.7 mm
Dimensions (LxW) 1 x 0.5 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Height (English) 0.5 in.
Height (Metric) 12.7 mm