Learn More
Brady™ Accessories for BMP™51 and BMP™53 Label Printers
Click to view available options
Product Type:
BMP50 Series Spare Battery Tray
Hard Case
Magnet Accessory
Unit Size:
Each
Description
- BMP™51 and BMP™53 Printer Accessories
- Printer Cases designed for use with BMP™51 and BMP™53 Label Printers from Brady™ protects your printer against impacts and dust while allowing for easy mobility
- Printer Magnet Attachment allows printer to affix to panel door or metal surface for more hands-free operation or storage
- Attaches to back of printer with screws to ensure stability
- Printer Battery Tray fits 8 AA batteries
- User may pre-load batteries into the spare tray for quick changeover in the field
- The battery tray snaps in place into the battery compartment
Specifications
Specifications
| Product Type | BMP50 Series Spare Battery Tray |
| Color | Gray |
| For Use With (Equipment) | BMP51 Label Printer, BMP53 Label Printer |
| Sterility | Non-sterile |
| Description | BMP50 Series Spare Battery Tray for 8 AA batteries |
| Quantity | 1 Ea. |
| Alternate Part Number | M50-BATT-TRAY |