missing translation for 'onlineSavingsMsg'
Learn More

BQ-788, MedChemExpress

Product Code. 16456908
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16456908

Brand: Medchem Express HY15894A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BQ-788 is a potent, selective ETB receptor antagonist with IC50 of 1.2 nM for inhibition of ET-1 binding to human Girardi heart cells, poorly inhibiting the binding to ETA receptors in human neuroblastoma cell line SK-N-MC cells with IC50 of 1300 nM.

BQ-788 is a potent, selective ETB receptor antagonist with IC50 of 1.2 nM for inhibition of ET-1 binding to human Girardi heart cells, poorly inhibiting the binding to ETA receptors in human neuroblastoma cell line SK-N-MC cells with IC50 of 1300 nM.

Specifications

Chemical Name or Material BQ-788
CAS 173326-37-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C34H51N5O7
Quantity 10 mg
Solubility Information DMSO : 170 mg/mL (264.88 mM; ultrasonic and warming and heat to 60°C)
SMILES CCCC[C@H](C(O)=O)NC([C@@H](CC1=CN(C(OC)=O)C2=CC=CC=C12)NC([C@H](CC(C)(C)C)NC(N3[C@H](C)CCC[C@@H]3C)=O)=O)=O
Molecular Weight (g/mol) 641.8
Formula Weight 641.8
Percent Purity 98.28%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.