Learn More
BPO-27 racemate, MedChemExpress
Product Code. 16460368
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460368 5 mg 5mg
16493847 50 mg 50mg
16467716 100 mg 100mg
16431869 1 mg 1mg
16427908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16460368 Supplier Medchem Express Supplier No. HY19778A
MedChemExpress BPO-27 racemate is a potent CFTR inhibitor with an IC50 of 8 nM.
BPO-27 racemate is a potent CFTR inhibitor with an IC50 of 8 nM.
Specifications
Chemical Name or Material BPO-27 racemate
CAS 1314873-02-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H18BrN3O6
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (30.40 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OC(C2=CC=C(Br)O2)C3=C(N(C)C4=O)C(C(N4C)=O)=C(C5=CC=CC=C5)N36)C6=C1)O
Molecular Weight (g/mol) 548.34
Formula Weight 548.34
Percent Purity 98.61%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus