Learn More
Trajan™ BP5 GC Capillary Column
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
Specifications
| Max. Temperature | 320/340°C |
| Diameter (Metric) Inner | 0.22 mm |
| Stationary Phase | 5% Phenyl/95% Dimethyl Polysiloxane |
| USP Type | G36, G41 |
| For Use With (Application) | Drugs of Abuse Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons |
| Temperature Range | -60°C to +320/340°C |
| Film Thickness | 0.25 μm |
| Length (Metric) | 25 m |
| Packing Material | Forte™ BP5 |
| Polarity | Non polar |
| Show More |