Learn More
Lonza™ Bovine Pituitary Extract (BPE), 13mg/mL, 2 mL
Product Code. 11685480
Change view
Click to view available options
Quantity:
2 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11685480 2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11685480 Supplier Lonza™ Supplier No. CC4009
Useful supplement in the culture of various epithelial cell types.
  • Suitable for use in the culture of various epithelial cell types.
  • The product is included in SingleQuots™ Kits associated with various media systems for epithelial cell types.
  • Available at a concentration of 13 mg/mL.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 13 mg/mL
Product Type Bovine Pituitary Extract
Quantity 2 mL