Learn More
Stuart™ Bottle Top Adapters
Product Code. 12457762
Click to view available options
Thread Size:
38 mm
40 mm
Unit Size:
Each
Product Code. 12457762
Brand: Stuart™ 1622383
Only null left
Add to basket
Add to basket
Stuart™ Bottle Top Adapters available in two different thread sizes
  • Available in various thread sizes
  • Polypropylene
  • 1 Adapter/Cs.
Specifications
Material Polypropylene
For Use With (Equipment) Liquid Handling Products
Quantity 1 Adapter/Cs.
Thread Size 38 mm