missing translation for 'onlineSavingsMsg'
Learn More

BOS-172722, MedChemExpress

Product Code. 16487297
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487297 5 mg 5mg
16455436 100 mg 100mg
16450887 50 mg 50mg
16434588 10 mg 10mg
16423146 25 mg 25mg
16421169 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16487297 Supplier Medchem Express Supplier No. HY112162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BOS-172722 is an inhibitor of monopolar spindle 1 (MPS1) checkpoint with an IC50 of 11 nM and 63 nM for MPS1 (1 mM ATP) and P-MPS1, respectively. BOS-172722 also has potential for the study of various forms of breast cancer.

BOS-172722 is an inhibitor of monopolar spindle 1 (MPS1) checkpoint with an IC50 of 11 nM and 63 nM for MPS1 (1 mM ATP) and P-MPS1, respectively. BOS-172722 also has potential for the study of various forms of breast cancer.

Specifications

Chemical Name or Material BOS-172722
CAS 1578245-44-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H30N8O
Quantity 5 mg
Solubility Information 1M HCl : 65 mg/mL (145.56 mM; ultrasonic and adjust pH to 2 with HCl) ∣DMSO : 5 mg/mL (11.20 mM; Need ultrasonic)
SMILES CC1=CC2=CN=C(NC3=CC=C(C4=NN=CN4C)C=C3OCC)N=C2C(NCC(C)(C)C)=N1
Molecular Weight (g/mol) 446.55
Formula Weight 446.55
Percent Purity 99.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.