Learn More
Bortezomib Trimer, TRC
Specifications
Specifications
| Chemical Name or Material | Bortezomib Trimer |
| CAS | 390800-88-1 |
| Molecular Formula | C57 H69 B3 N12 O9 |
| Quantity | 5 mg |
| Synonym | 2-Pyrazinecarboxamide, N,N',N''-[2,4,6-boroxintriyltris[[(1R)-3-methylbutylidene]imino[(1S)-2-oxo-1-(phenylmethyl)-2,1-ethanediyl]]]tris- (ACI), N,N',N''-[2,4,6-Boroxintriyltris[[(1R)-3-methylbutylidene]imino[(1S)-2-oxo-1-(phenylmethyl)-2,1-ethanediyl]]]t |
| InChI Formula | InChI=1S/C57H69B3N12O9/c1-37(2)28-49(70-52(73)43(31-40-16-10-7-11-17-40)67-55(76)46-34-61-22-25-64-46)58-79-59(50(29-38(3)4)71-53(74)44(32-41-18-12-8-13-19-41)68-56(77)47-35-62-23-26-65-47)81-60(80-58)51(30-39(5)6)72-54(75)45(33-42-20-14-9-15-21-42)69-57( |
| SMILES | CC(C)C[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)c2cnccn2)B3OB(OB(O3)[C@H](CC(C)C)NC(=O)[C@H](Cc4ccccc4)NC(=O)c5cnccn5)[C@H](CC(C)C)NC(=O)[C@H](Cc6ccccc6)NC(=O)c7cnccn7 |
| IUPAC Name | N-[(2S)-1-[[(1R)-1-[4,6-bis[(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]-1,3,5,2,4,6-trioxatriborinan-2-yl]-3-methylbutyl]amino]-1-oxo-3-phenylpropan-2-yl]pyrazine-2-carboxamide |
| Molecular Weight (g/mol) | 1098.666 |
| Formula Weight | 1098.559 |