Description
- Borosilicate glass (Boro 3.3) construction
- Graduation of approximate volume and inscriptions in high contrast white enamel
- Wall thickness approximately 1.2mm
- Without stopper
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass |
| Includes | High contrast white enamel markings |
| Packaging | 10Pack |
| Graduated | Yes |
| Product Type | Test Tube |