Learn More
Kimble Chase™ Glass Shell Vial with Closure
Product Code. 11873183
Click to view available options
Unit Size:
Pack of 144
Product Code. 11873183
Brand: Kimble Chase™ 60965D7
Only null left
Add to basket
Add to basket
Kimble Chase™ Glass Shell Vial with Closure features an open-bottom plug style plastic closure.
  • TITESEAL™ vials
  • Plain tops
  • Open-bottom plug style plastic closures, unattached
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
TRUSTED_SUSTAINABILITY
Specifications
No. per Case 432
No. per Pack 144
Closure Type Open Bottom Plug