Learn More
Kimble Chase™ Borosilicate Glass Shell Vials
Product Code. 10052582
Click to view available options
Capacity (Metric):
2 mL
Unit Size:
Pack of 1440
Product Code. 10052582
Brand: Kimble Chase™ 6093112
Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.
  • Straight-sided
  • Clear N-51A borosilicate glass
  • Without caps
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Capacity (Metric) 2 mL
Material Borosilicate Glass
For Use With (Application) Ideal for Storing Dry Products
Height (Metric) 35 mm
Type Shell Vial
Graduated Yes