Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with Solid Glass Plug
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 2
Description
- Cylindrical shape with straight discharge tube
- With NS socket and PE stopper
- With solid glass plug
- With Screwthreade retaining device
- With scale
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| Stopper Material | Polyethylene |