Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Joint Number:
19/26 NS
29/32 NS
Pore Size:
2.5mm
4mm
6mm
Unit Size:
Pack of 2
Description
- With NS socket and PE stopper
- Without scale, with PTFE plug
- With screwthread retaining device
- With inscription label
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass |
| Joint Number | 29/32 NS |
| Porosity | Yes |
| Pore Size | 2.5mm |
| Product Type | Separatory Funnel |