Learn More
Borosilicate Glass Petri Dishes
Constructed of borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Petri Dishes are autoclavable and include lid.
10.95 EUR - 200.00 EUR
Specifications
Closure Included | Closure Included |
---|---|
Closure Material | Borosilicate Glass |
Material | Borosilicate Glass |
Shape | Round |
Type | Petri Dish |
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
12921408
|
Fisherbrand
|
100 mm | 15 mm |
163.00 EUR
Pack of 18 |
In Stock
Log in to see stock. |
||||
12901408
|
Fisherbrand
|
60 mm | 12 mm |
52.30 EUR
Pack of 18 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
12931408
|
Fisherbrand
|
120 mm | 20 mm |
200.00 EUR
Pack of 18 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
12941408
|
Fisherbrand
|
150 mm | 25 mm |
10.95 EUR
Each |
In Stock
Log in to see stock. |
||||
12911408
|
Fisherbrand
|
80 mm | 15 mm |
80.10 EUR
Pack of 18 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
16190741
|
Fisherbrand
|
100 mm | 15 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16110751
|
Fisherbrand
|
150 mm | 25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16120751
|
Fisherbrand
|
60 mm | 12 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16130751
|
Fisherbrand
|
120 mm | 20 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Constructed of borosilicate glass.
- Includes lid
- Autoclavable
Specifications
Closure Included | |
Borosilicate Glass | |
Petri Dish |
Borosilicate Glass | |
Round |