Learn More
Fisherbrand™ Glass Square Coverslips
Product Code. 12343138 Shop All Fisherbrand Products
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
24 mm
Unit Size:
Pack of 200
Product Code. 12343138
Only null left
Add to basket
Add to basket
Glass Square Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product Type Coverslip
Shape Square
Material Soda Lime Glass
Length (Metric) 18 mm
Width (Metric) 18 mm
Thickness (Metric) #1.5 (0.16 to 0.19 mm)
Quantity 200/Pk.