Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc
Description
- Acc. to DIN 12596, complete with head
- Corrosion-free and resistant to thermal shock
- Available in a range of capacities
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | Borosilicate Glass |
| Type | Gas Washing Bottle with Filter Disc |
| Includes | Filter disc |
| Porosity | 1 |
| Standard Taper Joint | NS 29/32 |
| Certifications/Compliance | DIN 12596 |
| Quantity | 1 Pack |