Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
Click to view available options
Capacity (Metric):
50 mL
100 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Description
- Without scale
- With NS socket and PE stopper
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass |
| Stopper Material | Polyethylene |