Learn More
Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug
Product Code. 10333421
Click to view available options
Capacity (Metric):
50 mL
100 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. 10333421
Brand: Lenz Laborglasinstrumente™ 04004173
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels Acc. to Squibb, with PTFE Plug are manufactured from durable DURAN™ borosilicate glass.
  • Without scale
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Stopper Material Polyethylene