Learn More
IKA™ Borosilicate Glass Coated Receiving Flask
Shop All IKA Products
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
3000 mL
Unit Size:
Each
Specifications
Specifications
| Material | Borosilicate Glass |
| Type | Receiving Flask |
| Quantity | 1 Each |
| Volume (Metric) | 250 mL |