Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Centrifuge Tube
Product Code. 11894050 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
160 mm
167 mm
203 mm
Unit Size:
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11894050 167 mm Pack of 2
11874050 203 mm Pack of 2
11884050 160 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11894050 Supplier BRAND™ Supplier No. 362338
Only null left
Add to basket
Add to basket
Withstands RCF up to 700. Design and accuracy meet ASTM (American Society for Testing and Materials) specifications.
  • Withstands RCF up to 700.
  • Borosilicate glass construction (Boro 3.3)
  • Cylindrical with conical bottom
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
No. per Pack 2
Length (Metric) 167 mm
Bottom Shape Conical
Diameter (Metric) 96 mm
Product Type Centrifuge Tube
Max. RCF 700 x g