Learn More
BRAND™ Boro 3.3 Conical Ground Joint Stopper
Shop All Brand Products
Click to view available options
Standard Taper Joint:
NS 24/29
NS 45/40
Unit Size:
Each
Description
Specifications
Specifications
| Description | Solid |
| Standard Taper Joint | NS 24/29 |
| Material | Borosilicate Glass |
| Shape | Octagonal Grip |
Product Content Correction
Product Title
Spot an opportunity for improvement?Übermitteln Sie eine inhaltliche Korrektur