Learn More
Borax Anhydrous MP Biomedicals
Product Code. 11401841 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. 11401841
Brand: MP Biomedicals 0219030990
Chemical Identifiers
CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
Synonym Sodium Tetraborate, Sodium Borate Anhydrous, Sodium Borate
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Specifications
Quantity 500 g
Formula Weight 201.2
Density 2.367 g/mL
Chemical Name or Material Borax, Anhydrous