Learn More
Borane Ammonia Complex, TRC
Specifications
Specifications
| Chemical Name or Material | Borane Ammonia Complex |
| CAS | 13774-81-7 |
| Molecular Formula | NH3 . (BH3) |
| Quantity | 100 mg |
| Synonym | Ammineborane, Amminetrihydroboron, Ammonia Borane, Ammonia compd. with Borane (1:1), Ammonia, mixt. contg., Borane, mixt. with Ammonia, Borazane |
| InChI Formula | InChI=1S/BH3.H3N/h2*1H3 |
| SMILES | B.N |
| IUPAC Name | azane;borane |
| Molecular Weight (g/mol) | 30.87 |
| Formula Weight | 31.0593 |