Learn More
Emma Safety Footwear Bologna XD Safety Shoes
Click to view available options
Size:
39
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- S3-Safety shoe with sole plate for wet work
- Nitrile rubber heat resistant outsole
- Full grain leather construction
- ESD certified
- Chrome-free lining
- Sanitized
Specifications
Specifications
| Design | Bologna XD |
| Material | Leather |
| Size | 39 |