Learn More
Emma Safety Footwear Bologna Brogue-Style Business Shoes
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Polished black full-grain leather uppers
- Impenetrable nitrile rubber sole
- Hydro-Tec™ sanitized silver and wellness leather inlay sole
- Woven zero penetration insole
- Anti-bacterial with leather top layer in-lay sole
- Antistatic properties: ESD 0,1 - 35 M Ohm / NEN-EN-IEC 61340-5-1
Specifications
Specifications
| Color | Black |
| Material | Leather |
| Shank Type | Ankle support |
| Size | 47 |
| Sole Features | Anti-bacterial with leather top layer inlay sole |
| Sole Material | Impenetrable nitrile rubber |
| Style | Brogue |
| Certifications/Compliance (Europe) | EN ISO 20345:2011; ESD 0,1-35 M Ohm/NEN-EN-IEC 61340-5-1 |
| Product Type | Business Shoe |
Product Content Correction
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering