missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18017-1-AP-150UL
This item is not returnable.
View return policy
Description
CGI-143 encodes proteins that seem to be involved in cell proliferation or cell-cycle regulation, but the molecular function is still unknown.Specifications
| BOLA1 | |
| Polyclonal | |
| Unconjugated | |
| BOLA1 | |
| bolA homolog 1 (E. coli), BolA like protein 1, BOLA1, CGI 143, hBolA, RP11 196G18.18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 365875, 51027, 69168 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D8S9, Q9Y3E2 | |
| BOLA1 | |
| BOLA1 Fusion Protein Ag12565 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction