Learn More
Bohlender™ BOLA™ Screw-in Stopper
Description
- For connection to BOLA™ Reactor Lids
- For closure of non-used connectors
- Male thread
- With hexagonal gripping surface in standard wrench size
Specifications
Specifications
| Type | Hexagonal Grip |
| Material | PTFE |
| Joint Type | Screw-in Thread NPT 1/4 |
| Size | Wrench Size (SW) 15 mm |