Learn More
Bohlender™ BOLA™ Screw-in Stopper
Product Code. 17106954
Change view
Click to view available options
Joint Type:
Screw-in Thread NPT 1
Screw-in Thread NPT 1/2
Screw-in Thread NPT 1/4
Screw-in Thread NPT 3/4
Size:
Wrench Size (SW) 15 mm
Wrench Size (SW) 22 mm
Wrench Size (SW) 32 mm
Wrench Size (SW) 45 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Type Size unitSize
17106954 Screw-in Thread NPT 1/4 Wrench Size (SW) 15 mm Each
17116954 Screw-in Thread NPT 1/2 Wrench Size (SW) 22 mm Each
17126954 Screw-in Thread NPT 3/4 Wrench Size (SW) 32 mm Each
17136954 Screw-in Thread NPT 1 Wrench Size (SW) 45 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17106954 Supplier Bohlender™ Supplier No. B15302
Only null left
Add to basket
Add to basket
Screw-in stopper with male thread and hexagonal grip for Reactor Lids.
  • For connection to BOLA™ Reactor Lids
  • For closure of non-used connectors
  • Male thread
  • With hexagonal gripping surface in standard wrench size
Specifications
Type Hexagonal Grip
Material PTFE
Joint Type Screw-in Thread NPT 1/4
Size Wrench Size (SW) 15 mm