Learn More
Bohlender™ BOLA™ Reactor Lids PTFE-EX
Product Code. 17196954
Click to view available options
Type:
4 x NPT Female Threads
6x NPT Female Threads
Unit Size:
Each
Product Code. 17196954
Brand: Bohlender™ B17004
Reactor lids made of PTFE-EX suitable for glass reactors with Schott flat flange.
  • Made of static dissipative PTFE-EX
  • Suitable for glass reactors DN 100 with SCHOTT flat flange
  • With screw-in threads for connection of transition fittings
  • Bore with female thread M5x6 for earthing connection on the upper side of the lid
  • Easy assembly
  • Flexibly expandable
  • Compatible with glass reactors with SCHOTT-flat flange
  • Universal chemical resistance
  • With connectors for the use of existing equipment with ground joint or GL thread
Thread connections in detail:
  • Centric thread connection (purple) for insertion of a stirrer bearing
  • Vertical thread connection (blue) with parallel alignment to the stirrer shaft
  • Inclined thread connection (yellow) with direction straight to the stirrer shaft
  • Inclined thread connection (green) with direction aside the stirrer shaft
Specifications
Includes Center Neck 1/2 in., Lateral Necks 3 x 1/2 in.
Type 4 x NPT Female Threads
Material PTFE-EX
For Use With (Equipment) Glass Reactors DN 60 with SCHOTT Flat Flange
Hose Connection No. NPT 1/2 in. aside the shaft