Learn More
Bohlender™ BOLA™ Reactor Lids PTFE-EX
Click to view available options
Type:
4 x NPT Female Threads
6x NPT Female Threads
Unit Size:
Each
Description
- Made of static dissipative PTFE-EX
- Suitable for glass reactors DN 100 with SCHOTT flat flange
- With screw-in threads for connection of transition fittings
- Bore with female thread M5x6 for earthing connection on the upper side of the lid
- Easy assembly
- Flexibly expandable
- Compatible with glass reactors with SCHOTT-flat flange
- Universal chemical resistance
- With connectors for the use of existing equipment with ground joint or GL thread
Thread connections in detail:
- Centric thread connection (purple) for insertion of a stirrer bearing
- Vertical thread connection (blue) with parallel alignment to the stirrer shaft
- Inclined thread connection (yellow) with direction straight to the stirrer shaft
- Inclined thread connection (green) with direction aside the stirrer shaft
Specifications
Specifications
| Includes | Center Neck 1/2 in., Lateral Necks 3 x 1/2 in. |
| Type | 4 x NPT Female Threads |
| Material | PTFE-EX |
| For Use With (Equipment) | Glass Reactors DN 60 with SCHOTT Flat Flange |
| Hose Connection No. | NPT 1/2 in. aside the shaft |