Learn More
Bohlender™ BOLA™ Flat Flange Lids
Product Code. 17136964
Click to view available options
Joint Type:
DN 100
DN 150
DN 60
Standard Taper Joint:
NS 14/23, GL 18
NS 29/32
NS 29/32, NS 19/26, GL 25
Unit Size:
Each
Product Code. 17136964
Brand: Bohlender™ B21002
Flat Flange Lids suitable for reaction vessels with SCHOTT flat flange.
  • Suitable for reaction vessels with SCHOTT flat flange
  • Center neck socket NS 29/32
  • Lateral necks with ground joint sockets or GL-thread
  • Bottom side with centring collar for non-slipping alignment on the vessel
  • All lateral connectors are made to insert tubing, tubes and probes either aside the center or vertical (parallel to the stirrer shaft) in order to avoid collisions
Specifications
Type Flat Flange Lid
Material PTFE
For Use With (Equipment) Reaction Vessels with SCHOTT Flat Flange
Standard Taper Joint NS 14/23, GL 18
Joint Type DN 60
Size Lateral Necks: 2 x NS 14/23 and 2 x GL 18 Angled Aside the Shaft