Learn More
Bohlender™ BOLA™ Double Tube End Fittings
Description
- Made of PTFE
- Two male threads UNF 1/4 in. 28 G
- Packing unit: 10 pieces; differing ordering quantities are rounded up to factor of 10
Specifications
Specifications
| Type | Fitting |
| Color | White |
| Packaging Quantity | 10 Pk. |
| Thread Size | UNF 1/4 in. 28 G |
| For Use With | Tubing (I.D. x O.D.): 1.6 x 3.2 mm |
| Material | PTFE |