Learn More
WTW™ BOD Test Kits
Product Code. 15381437
Change view
Click to view available options
Range:
0.5 to 3000 mg/L BOD
Unit Size:
1 set
Product Code. Range unitSize
15381437 0.5 to 3000 mg/L BOD 1 set
1 options
Product Code. 15381437
Brand: WTW™ 252028
Obtain precision photometric measurements for waste water, drinking water, and surface water with WTW™ BOD test Kits.
  • For wastewater, production control, ground water, surface water
  • For use with PhotoLab™ models S6, S12 and 6000/7000
  • Biological Oxygen Demand (BOD) Test Kits
  • This test is highly accurate and easy to use
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PhotoLab™ Photometers
No. of Tests 50
Range 0.5 to 3000 mg/L BOD
Test Method Photometric
compliance-icons compliance-icons compliance-icons
Product Identifier
  • BOD Cell Test Method: photometric 0.5 - 3000 mg/l
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.