missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-MLF TFA, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Boc-MLF (TFA) is a peptide, used as a specific formyl peptide receptor (FPR) antagonist, also inhibits the signaling through formyl peptide receptor like 1 (FPRL1) at higher concentrations.
Specifications
Specifications
| Chemical Name or Material | Boc-MLF TFA |
| Physical Form | Solid |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C27H40F3N3O8S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Boc-Met-Leu-Phe-OH (TFA) |
| Solubility Information | DMSO : 150 mg/mL (240.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(O)[C@H](CC1=CC=CC=C1)NC([C@H](CC(C)C)NC([C@H](CCSC)NC(OC(C)(C)C)=O)=O)=O.OC(C(F)(F)F)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction