missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMS-986158, MedChemExpress
Description
BMS-986158 is a potent BET inhibitor with IC50s of 6.6 and 5 nM in NCI-H211 small cell lung cancer (SCLC) cells and MDA-MB231 triple negative breast cancer (TNBC) cells, respectively.
Specifications
Specifications
| Chemical Name or Material | BMS-986158 |
| CAS | 1800340-40-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H33N5O2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (100.88 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | OC(C)(C)C1=CC=C2C(N([C@H](C3=CC=CC=C3)C4CCOCC4)C5=C2N=CC(C6=C(C)N=NN6C)=C5)=C1 |
| Molecular Weight (g/mol) | 495.62 |
| Formula Weight | 495.62 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?