missing translation for 'onlineSavingsMsg'
Learn More

BMS-833923, MedChemExpress

Product Code. p-200100984
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495868

Brand: Medchem Express HY13809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-833923 (XL-139) is an orally bioavailable small-molecule inhibitor of Smoothened with potential antineoplastic activity; inhibits BODIPY cyclopamine binding to SMO in a dose-dependent manner with an IC50 of 21 nM.

BMS-833923 (XL-139) is an orally bioavailable small-molecule inhibitor of Smoothened with potential antineoplastic activity

Specifications

Chemical Name or Material BMS-833923
CAS 1059734-66-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H27N5O
Quantity 10 mg
Synonym XL-139
Solubility Information DMSO : 50 mg/mL (105.58 mM; Need ultrasonic)
SMILES O=C(NC1=CC(CNC)=CC=C1C)C2=CC=C(NC3=NC(C4=CC=CC=C4)=C5C=CC=CC5=N3)C=C2
Molecular Weight (g/mol) 473.57
Formula Weight 473.57
Percent Purity 98.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.