missing translation for 'onlineSavingsMsg'
Learn More

BMS-303141, MedChemExpress

Product Code. 16426988
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426988 10 mg 10mg
16493037 50 mg 50mg
16479527 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16426988 Supplier Medchem Express Supplier No. HY16107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BMS-303141 is a potent, cell-permeable ATP-citrate lyase (ACL) inhibitor with an IC50 of 0.13 μM.

BMS-303141 is a potent, cell-permeable ATP-citrate lyase (ACL) inhibitor with an IC50 of 0.13 μM.

Specifications

Chemical Name or Material BMS-303141
CAS 943962-47-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H15Cl2NO4S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (58.92 mM; Need ultrasonic)
SMILES O=S(C1=CC(Cl)=CC(Cl)=C1O)(NC2=CC(C3=CC=CC=C3)=CC=C2OC)=O
Molecular Weight (g/mol) 424.3
Formula Weight 424.3
Percent Purity 98.47%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.