missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP-5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15558312
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15558312 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558312 Supplier Invitrogen™ Supplier No. PA547821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

In Western blots, approximately 20% cross-reactivity with recombinant human BMP-5 is observed. In direct ELISAs, approximately 100% cross-reactivity with recombinant human BMP-5 is observed, and approximately 4% cross-reactivity with recombinant mouse (rm) BMP-6 and rmBMP-7 is observed, and less than 1% cross-reactivity with rmBMP-8b is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth, and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling, and maintenance of a variety of other tissues and organs. BMP-5 is expressed in the nervous system, lungs and liver. It is a known regulator for dendritic growth in sympathetic neurons. BMP-5 is a 454 amino acid precursor protein that is cleaved to release the biologically active C-terminal mature protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMP-5
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Bmp5
Gene Accession No. P49003
Gene Alias AU023399; BMP; BMP 5; Bmp5; BMP-5; bone morphogenenic protein-5; Bone morphogenetic protein; bone morphogenetic protein 5; MGC34244; se; short ear
Gene Symbols Bmp5
Host Species Sheep
Immunogen CHO-derived recombinant mouse BMP-5 Ala315-His452.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12160
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.