Learn More
SI Analytics™ BlueLine Plug Cable Combination
Shop All SI Analytics Products
Click to view available options
Connector Types:
BNC
DIN
Length (Metric) Cable:
1 m
3 m
Unit Size:
Each
Description
- For use with BlueLine series
- Coaxial cable
- Available in 1m or 3m lengths
Specifications
Specifications
| Connector Types | BNC |
| For Use With (Equipment) | BlueLine |
| Type | LB 1 BNC |
| Includes | 1m Coaxial Cable, BNC Instrument Plug |
| Length (Metric) Cable | 1 m |