Learn More
3M™ Blue SMS Based Polypropylene Category III Protective Coveralls
Click to view available options
Unit Size:
Pack of 20
Specifications
Specifications
| Product Type | Protective Coveralls |
| Waist Style | Elastic |
| Neck Style | Hooded |
| Hood Type | 3-panel |
| Protection Class | Category III |
| Resistance | Flame |
| Height (English) | 68.5 to 71.3 in. |
| Height (Metric) | 174 to 181 cm |
| Style | Enhanced Gusset |