Learn More
Hirschmann™ Class B Glass Burettes with Straight Glass Stopcock, Blue Graduation
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 2
Description
- Schellbach
- Short line
- With blue graduation
- Acc. to DIN EN ISO 385
- Straight stopcock with glass key
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Glass |
| Graduations | 0.1 mL |
| Includes | Schellbach Stripes |
| Scale | Blue |
| Stopcock | Straight |
| Stopcock Material | Glass |
| Tolerance | ±0.075 mL |
| Type | Buret |
| Class | Class B |
| Show More |