Learn More
Hirschmann™ Class AS Burettes Straight Glass Stopcock with PTFE Key, Blue Graduation
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 2
Description
- Schellbach
- Main point ring graduation
- With blue graduation
- Acc. to DIN EN ISO 385
- Straight stopcock with PTFE key
- Conformity certified, with dated batch identification
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Glass |
| Graduations | 0.1 mL |
| Includes | Schellbach Stripes |
| Scale | Blue |
| Stopcock | Straight |
| Stopcock Material | PTFE |
| Tolerance | ±0.05 mL |
| Type | Buret |
| Class | Class AS |
| Show More |