Learn More
Invitrogen™ BLOCK-iT™ Transfection Optimization Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The BLOCK-iT™ Transfection Optimization Kit is designed to optimize RNAi transfection using controls for transfection and viability. Each kit contains a BLOCK-iT™ Fluorescent Oligo, which provides:
A strong, easily detectable FITC signal indicating transfection efficiency
Chemical modifications for a clearer, more persistent cellular signal than labeled siRNA provides
Proven correlation of transfection efficiency with Stealth RNAi™ siRNA and traditional siRNA molecules
This makes BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. In addition, the kit contains a Positive Control Stealth RNAi™ siRNA molecule directed at human p53 and a Scrambled Stealth RNAi™ siRNA molecule as a negative control. To assay cell viability, Dead Cell Dye is included. This dye colors dead cells as red, thereby allowing easy identification of non-viable cells in a tissue culture plate. Using these reagents together allows you to both optimize transfection conditions and have a functional positive control in every RNAi experiment you perform.
This makes BLOCK-iT™ Fluorescent Oligo an ideal indicator of transfection efficiency for RNAi experiments. In addition, the kit contains a Positive Control Stealth RNAi™ siRNA molecule directed at human p53 and a Scrambled Stealth RNAi™ siRNA molecule as a negative control. To assay cell viability, Dead Cell Dye is included. This dye colors dead cells as red, thereby allowing easy identification of non-viable cells in a tissue culture plate. Using these reagents together allows you to both optimize transfection conditions and have a functional positive control in every RNAi experiment you perform.
Specifications
Specifications
| Control Type | Cell Viability Control, Negative Control, Transfection Control, Positive Control |
| Format | Kit |
| Sample Type | RNA |
| Vector | pLATE |
| For Use With (Application) | Transfection |
| Product Line | BLOCK-iT™, Stealth RNAi™ siRNA |
| Product Type | Transfection Kit |
| Quantity | 1 Kit |
| RNAi Type | siRNA |