Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Calibrated Density Bottles
Calibrated density bottle made with Borosilicate glass that is ideal for use in most research, scientific, and other types of laboratory environments.
Brand:  BRAND™ 43328
66.52 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.05400kg
 View more versions of this product
Product Code. 10748171
  • 75.00 EUR / Each
Estimated Shipment: 25-06-2024
Description
  • Calibrated density bottle made with Borosilicate glass
  • DIN ISO 3507, type Gay-Lussac
  • Calibrated to contain (TC, In)
  • Includes individual certificate
  • Stopper NS 10/19 with capillary
  • Top of stopper ground and polished
  • The volume in cm³ is specified to a precision of 3 decimal places
Specifications
Specifications
1Pack
50 mL
Borosilicate Glass
Calibrated Density Bottle