Learn More
BRAND™ Blaubrand™ Borosilicate Glass Class A USP Volumetric Flask
Product Code. p-8000977 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Closure Size:
10/19
12/21
14/23
19/26
24/29
29/32
Unit Size:
Each
Pack of 2
Product Code. 11814180
Brand: BRAND™ 36938
Only null left
Add to basket
Add to basket
Manufactured from Class A Borosilicate Glass and complies with DIN EN ISO 1042 standards.BRAND™ Blaubrand™ Borosilicate Glass Class A USP Volumetric Flask includes polypropylene stopper and USP batch certificate. DE-M Marking provides easy volume check.
  • Manufactured from Class A Borosilicate Glass.
  • Complies with DIN EN ISO 1042 standards.
  • Polypropylene stopper included.
  • DE-M Marking provides easy volume check.
  • Calibrated to contain. (TC, In)
  • USP batch certificate included. USP individual certificate available on request.
TRUSTED_SUSTAINABILITY
Specifications
Calibration Calibrated To Contain
Certifications/Compliance DIN EN ISO 1042, USP batch certificate, USP individual certificate upon request
Closure Material Polypropylene
Color Clear
Capacity (Metric) 5 mL
Material Borosilicate Glass
Limit of Error ±0.020 mL
No. per Case 2 Piece
Class A
Closure Size 10/19