Learn More
Brady™ Black 7964 Series Thermal Transfer Printer Ribbon
Provides good chemical resistance to solvents and sharp printing
Brand:  Brady™ 237588
Additional Details : Weight : 0.10900kg
 View more versions of this product
Product Code. 11523165
  • 43.75 EUR / 1 box
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • Compatible with various Brady labels
Specifications
Specifications
Black
110 mm
Black 7964 Series Thermal Transfer Printer Ribbon
1 Ea.
100 m
12.7 mm I.D.
Ink side in