missing translation for 'onlineSavingsMsg'
Learn More

BIX 02565, MedChemExpress

Product Code. 16496978
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496978 10 mg 10mg
16467056 100 mg 100mg
16463037 50 mg 50mg
16449527 5 mg 5mg
16430276 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496978 Supplier Medchem Express Supplier No. HY16104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BIX 02565 is a potent ribosomal S6 kinase 2 (RSK2) inhibitor with IC50 of 1.1 nM.

BIX 02565 is a potent ribosomal S6 kinase 2 (RSK2) inhibitor with IC50 of 1.1 nM.

Specifications

Chemical Name or Material BIX 02565
CAS 1311367-27-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H30N6O2
Quantity 10 mg
Solubility Information DMSO : 20.75 mg/mL (45.25 mM; Need ultrasonic and warming)
SMILES O=C(C1=CC2=C(C=C1)C=C3N2[C@H](C)CCNC3=O)NC4=NC5=CC=CC=C5N4CCCN(C)C
Molecular Weight (g/mol) 458.56
Formula Weight 458.56
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.