missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol F, TRC

Product Code. 30008966
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008966 1 g 1g
30136014 5 g 5g
30006767 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008966 Supplier Toronto Research Chemicals Supplier No. TRCB5195551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-92-8
Formula Weight 200.0837
InChI Formula InChI=1S/C13H12O2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8,14-15H,9H2
IUPAC Name 4-[(4-hydroxyphenyl)methyl]phenol
Molecular Formula C13 H12 O2
Molecular Weight (g/mol) 200.23
Percent Purity >95%
SMILES Oc1ccc(Cc2ccc(O)cc2)cc1
Synonym Phenol, 4,4'-methylenedi- (8CI), Phenol, p,p'-methylenedi- (4CI), 4,4'-Methylenebis[phenol], 1,1-Bis(4-hydroxyphenyl)methane, 4,4'-Bis(hydroxyphenyl)methane, 4,4'-Dihydroxydiphenylmethane, 4,4'-Methylenediphenol, Bis(4-hydroxyphenyl)methane, Bis(p-hydroxyphenyl)methane, Bisphenol F, HDM, NSC 401136, PP-BIP-F, p,p'-BPF, p-(p-Hydroxybenzyl)phenol
Chemical Name or Material Bisphenol F
Grade HPLC
Show More Show Less

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.