Learn More
Bisindolylmaleimide VIII Acetic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Bisindolylmaleimide VIII Acetic Acid Salt |
| CAS | 138516-31-1 |
| Molecular Formula | C26H26N4O4 |
| Quantity | 100 mg |
| Synonym | 3-[1-(3-Aminopropyl)-1H-indol-3-yl]-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione Acetic Acid Salt, Ro 31-7549 Acetic Acid Salt, Ro 31-7549/001 Acetic Acid Salt |
| InChI Formula | InChI=1S/C24H22N4O2.C2H4O2/c1-27-13-17(15-7-2-4-9-19(15)27)21-22(24(30)26-23(21)29)18-14-28(12-6-11-25)20-10-5-3-8-16(18)20;1-2(3)4/h2-5,7-10,13-14H,6,11-12,25H2,1H3,(H,26,29,30);1H3,(H,3,4) |
| SMILES | O=C(N1[H])C(C2=CN(C)C3=C2C=CC=C3)=C(C4=CN(CCCN)C5=C4C=CC=C5)C1=O.CC(O)=O |
| IUPAC Name | acetic acid;3-[1-(3-aminopropyl)indol-3-yl]-4-(1-methylindol-3-yl)pyrrole-2,5-dione |
| Molecular Weight (g/mol) | 458.51 |
| Formula Weight | 458.1954 |